Merck
Microbial degradation of four crude oil by biosurfactant producing strain Rhodococcus sp.
Bioresource technology (2017-02-27)
Yongrui Pi, Bing Chen, Mutai Bao, Fuqiang Fan, Qinhong Cai, Lv Ze, Baiyu Zhang
ZUSAMMENFASSUNG
Rhodococcus erythropolis M-25, one of the representative biosurfactant producers, performed effectively during the biodegradation of four crude oil. The microbial degradation efficiency is positively relevant to the API of the crude oil. The chemical dispersant Corexit 9500A did not enhance the biodegradation of the petroleum hydrocarbons during the experimental period. 70.7% of the N-4 oil was degraded after 30days, while in the Corexit 9500A plus sample the biodegradation removal was 42.8%. The Corexit-derived compounds were metabolized by M-25 at the same time of the petroleum hydrocarbons biodegrading. Neither biodegradation nor chemical dispersion process has almost no effect on the biomarker (m/z=231). The saturated methyl-branched fatty acids increased from 37.3%, to 49.4%, when M-25 was exposed with the N-4 crude oil. Similarly, the saturated methyl-branched fatty acids in the membrane of N3-2P increased from 20.25% to 44.1%, when exposed it with the N-4 crude oil.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
O-Acetyl-L-Carnitin -hydrochlorid, ≥99% (titration), powder