Merck
  • High-efficiency extracellular release of free fatty acids from Aspergillus oryzae using non-ionic surfactants.
High-efficiency extracellular release of free fatty acids from Aspergillus oryzae using non-ionic surfactants.
Journal of biotechnology (2017-03-17)
Koichi Tamano, Ai Miura, Hideaki Koike, Yasushi Kamisaka, Myco Umemura, Masayuki Machida
ZUSAMMENFASSUNG
Free fatty acids (FFAs) are useful for generating biofuel compounds and functional lipids. Microbes are increasingly exploited to produce FFAs via metabolic engineering. However, in many microorganisms, FFAs accumulate in the cytosol, and disrupting cells to extract them is energy intensive. Thus, a simple cost-effective extraction technique must be developed to remove this drawback. We found that FFAs were released from cells of the filamentous fungus Aspergillus oryzae with high efficiency when they were cultured or incubated with non-ionic surfactants such as Triton X-100. The surfactants did not reduce hyphal growth, even at 5% (w/v). When the faaA disruptant was cultured with 1% Triton X-100, more than 80% of the FFAs synthesized de novo were released. When the disruptant cells grown without surfactants were incubated for 1h in 1% Triton X-100 solution, more than 50% of the FFAs synthesized de novo were also released. Other non-ionic surfactants in the same ether series, such as Brij 58, IGEPAL CA-630, and Tergitol NP-40, elicited a similar FFA release. The dry cell weight of total hyphae decreased when grown with 1% Triton X-100. The decrement was 4.9-fold greater than the weight of the released FFAs, implying release of other intracellular compounds. Analysis of the culture supernatant showed that intracellular lactate dehydrogenase was also released, suggesting that FFAs are not released by a specific transporter. Therefore, ether-type non-ionic surfactants probably cause non-specific release of FFAs and other intracellular compounds by increasing cell membrane permeability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Brij® 58, average Mn ~1124