Merck
Triterpene derivatives from Euphorbia enhance resistance against Verticillium wilt of tomato.
Phytochemistry (2016-12-29)
Amal Smaili, Noureddine Mazoir, Lalla Aicha Rifai, Tayeb Koussa, Kacem Makroum, El Mostafa Kabil, Ahmed Benharref, Mohamed Faize
ZUSAMMENFASSUNG
Oxidation of α-euphorbol and 31-norlanostenol, two triterpenic compounds isolated from the latex of Euphorbia resinifera and Euphorbia officinarum respectively, yielded four products named 3β-tosyloxy-4α,14α-dimethyl-5α-cholesta-7,9-diene; 4α,14α-dimethyl-5α-cholesta-7,9-dien-3β-ol; 24-methylen-elemo-lanosta-8,24-dien-3-one and elemo-lanost-8-en-3,11,24-trione. They were evaluated for protection of tomato plants against Verticillium dahliae in a greenhouse. The four semisynthesized products were phytotoxic at higher concentrations as they completely inhibited tomato germination at 100 and 500 μg/ml. However at lower concentrations (10 and 50 μg/ml) germination and root length were not affected. Disease resistance against Verticillium wilt was assessed in tomato plants derived from seeds that germinated in the presence of 10 and 50 μg/ml of the four products. All of them were able to reduce significantly disease severity, with 10 μg/ml being more effective than 50 μg/ml. Reduction of leaf alteration index and of stunting index ranged from 52 to 68% and from 43 to 67%, respectively, while vessel discoloration was reduced by at least 95%. The compounds were also able to elicit H
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Chlorperbenzoesäure, ≤77%
Sigma-Aldrich
Ruthenium(III)-chlorid, Anhydrous, solid, Ru content 45-55%
Sigma-Aldrich
1,3-Propylene sulfite, 99%