- Peroxynitrite formed during a transient episode of brain ischaemia increases endothelium-derived hyperpolarization-type dilations in thromboxane/prostaglandin receptor-stimulated rat cerebral arteries.
Peroxynitrite formed during a transient episode of brain ischaemia increases endothelium-derived hyperpolarization-type dilations in thromboxane/prostaglandin receptor-stimulated rat cerebral arteries.
Acta physiologica (Oxford, England) (2016-09-30)
Y Onetti, A P Dantas, B Pérez, A J McNeish, E Vila, F Jiménez-Altayó
PMID27683007
ZUSAMMENFASSUNG
Increased thromboxane A Vascular function was determined by wire myography after TP stimulation with the thromboxane A In U46619-pre-constricted MCA, EDH-type relaxation by the proteinase-activated receptor 2 agonist serine-leucine-isoleucine-glycine-arginine-leucine-NH Following TP stimulation in MCA, EDH-type relaxation to SLIGRL is greater after I/R due to endothelial filamentous-actin disruption by peroxynitrite, which prevents TP-induced block of SK