Merck
  • Sensitive inexpensive spectrophotometric and spectrofluorimetric analysis of ezogabine, levetiracetam and topiramate in tablet formulations using Hantzsch condensation reaction.
Sensitive inexpensive spectrophotometric and spectrofluorimetric analysis of ezogabine, levetiracetam and topiramate in tablet formulations using Hantzsch condensation reaction.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-05-10)
F A Ibrahim, A F El-Yazbi, M M Wagih, M A Barary
ZUSAMMENFASSUNG
Two highly sensitive, simple and selective spectrophotometric and spectrofluorimetric assays have been investigated for the analysis of ezogabine, levetiracetam and topiramate in their pure and in pharmaceutical dosage forms. The suggested methods depend on the condensation of the primary amino-groups in the three drugs with acetylacetone and formaldehyde according to Hantzsch reaction yielding highly fluorescent yellow colored dihydropyridine derivatives. The reaction products of ezogabine, levetiracetam and topiramate were measured spectrophotometrically at 418, 390 and 380nm or spectrofluorimetrically at λ
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)