Merck
Co-occurring Mutations of Tumor Suppressor Genes,
Clinical cancer research : an official journal of the American Association for Cancer Research (2016-12-23)
Robin Tranchant, Lisa Quetel, Anne Tallet, Clement Meiller, Annie Renier, Leanne de Koning, Aurelien de Reynies, Francoise Le Pimpec-Barthes, Jessica Zucman-Rossi, Marie-Claude Jaurand, Didier Jean
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)