Merck
  • Effects of Hydrogen Sulfide-Releasing Compounds on Aqueous Humor Outflow Facility in Porcine Ocular Anterior Segments, Ex Vivo.
Effects of Hydrogen Sulfide-Releasing Compounds on Aqueous Humor Outflow Facility in Porcine Ocular Anterior Segments, Ex Vivo.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics (2017-01-19)
Jenaye Robinson, Esther Okoro, Chinoso Ezuedu, Leah Bush, Catherine A Opere, Sunny E Ohia, Ya Fatou Njie-Mbye
ZUSAMMENFASSUNG
To investigate the pharmacological actions of hydrogen sulfide (H Porcine ocular anterior segments were perfused with Dulbecco's modified Eagle's medium at a constant pressure of 7.35 mmHg. After stable outflow baseline, explants were exposed to NaHS or l-cysteine. The increase in outflow generated by the H l-cysteine elicited a concentration-dependent increase in AH outflow facility, reaching maximal effect at 100 nM (150.6% ± 17.2% of basal level). This increase in outflow induced by l-cysteine was significantly (P < 0.001) antagonized by AOAA (30 μM) and glibenclamide (100 μM). AOAA and glibenclamide had no significant action on baseline outflow, whereas SQ 22536 (100 μM) increased outflow for only an hour. In addition, NaHS produced a concentration-dependent increase in AH outflow, with a maximal effect at 10 μM (151.4% ± 22.9% of basal level). Likewise, the increase in outflow caused by NaHS was significantly (P < 0.04) blocked by glibenclamide and SQ 22536. H&E staining revealed that l-cysteine or NaHS did not alter TM conformation. H