Merck
Formetanate toxicity and changes in antioxidant enzyme system of Apis mellifera larvae.
Environmental science and pollution research international (2017-04-15)
Martin Staroň, Rastislav Sabo, Anna Sobeková, Lucia Sabová, Jaroslav Legáth, Ľuboslava Lohajová, Peter Javorský
ZUSAMMENFASSUNG
Substantial percentage of world food production depends on pollinating service of honeybees that directly depends on their health status. Among other factors, the success of bee colonies depends on health of developed larvae. The crucial phase of larval development is the first 6 days after hatching when a worker larva grows exponentially and larvae are potentially exposed to xenobiotics via diet. In the present study, we determined the lethal concentration LC