- Sensory evaluation of boar-taint-containing minced meat, dry-cured ham and dry fermented sausage by a trained expert panel and consumers.
Sensory evaluation of boar-taint-containing minced meat, dry-cured ham and dry fermented sausage by a trained expert panel and consumers.
Food chemistry (2017-05-23)
Kaat Verplanken, Jella Wauters, Vicky Vercruysse, Marijke Aluwé, Lynn Vanhaecke
PMID28530572
ZUSAMMENFASSUNG
One of the main issues related to entire male pigs is the occurrence of boar taint, an off-odour, which compromises meat consumability. In this study, odour thresholds (indole: 24-65µgkg
MATERIALIEN