Merck
Bacteriocin-Producing
Frontiers in microbiology (2017-06-03)
Allison Vimont, Benoît Fernandez, Riadh Hammami, Ahlem Ababsa, Hocine Daba, Ismaïl Fliss
ZUSAMMENFASSUNG
Bacterial isolates from raw camel milk were screened for antibacterial activity using the agar diffusion assay. Ten isolates selected for their inhibition of Gram-positive bacteria were identified by 16S sequencing as