Merck
  • Identification of an acidic endo-polygalacturonase from Penicillium oxalicum CZ1028 and its broad use in major tropical and subtropical fruit juices production.
Identification of an acidic endo-polygalacturonase from Penicillium oxalicum CZ1028 and its broad use in major tropical and subtropical fruit juices production.
Journal of bioscience and bioengineering (2017-03-04)
Zhong Cheng, Dong Chen, Qingyan Wang, Liang Xian, Bo Lu, Yutuo Wei, Hongchi Tang, Zhilong Lu, Qixia Zhu, Yunlai Chen, Ribo Huang
ZUSAMMENFASSUNG
Endo-polygalacturonases play an important role on depectinization in fruit juices industry. A putative endo-polygalacturonase gene PoxaEnPG28A was cloned from Penicillium oxalicum CZ1028. PoxaEnPG28A consisted of a putative signal peptide and a catalytic domain belonging to glycoside hydrolase family 28, and it shared 72% identity with that of a functionally characterized endo-polygalacturonase from Trichoderma harzianum. Gene PoxaEnPG28A was successfully expressed in Pichia pastoris with a high yield of 1828.7 U/mL. The purified recombinant enzyme PoxaEnPG28A hydrolyzed polygalacturonic acid in endo-manner releasing oligo-galacturonates. PoxaEnPG28A showed maximal activity at pH 5.5 and 55°C, and was stable between pH 3.0 to 10.0 and below 45°C. The kinetic constants K
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trigalacturonic acid, ≥90% (HPLC)