Merck
  • The Protective Effects of Helix B Surface Peptide on Experimental Acute Liver Injury Induced by Carbon Tetrachloride.
The Protective Effects of Helix B Surface Peptide on Experimental Acute Liver Injury Induced by Carbon Tetrachloride.
Digestive diseases and sciences (2017-04-04)
Shengdi Wu, Cheng Yang, Nuo Xu, Lingyan Wang, Yun Liu, Jiyao Wang, Xizhong Shen
ZUSAMMENFASSUNG
To investigate the protective effects of helix B surface peptide (HBSP) on acute liver injury induced by carbon tetrachloride (CCl HBSP (8 nmol/kg) was intraperitoneally injected into C57 BL/6 mice 2 h after CCl HBSP significantly decreased serum alanine aminotransferase, aspartate aminotransferase, lactate dehydrogenase, and pro-inflammatory cytokines in liver tissues after CCl HBSP is a potential therapeutic agent against acute liver injury induced by CCl
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
9-Ethylcarbazol, 97%