Merck
Asenapine modulates mood-related behaviors and 5-HT
CNS neuroscience & therapeutics (2017-04-19)
Sarah Delcourte, Erika Abrial, Adeline Etiévant, Renaud Rovera, Jørn Arnt, Michael Didriksen, Nasser Haddjeri
ZUSAMMENFASSUNG
Asenapine is a new atypical antipsychotic prescribed for the treatment of psychosis/bipolar disorders that presents higher affinity for serotonergic than dopaminergic receptors. The objective of this study was to investigate its antidepressant-like and antimanic-like properties on relevant animal models of depression and mania and to assess the acute and chronic effect of Asenapine on dorsal raphe nucleus (DRN) 5-HT cell firing activity. We assessed the effects of Asenapine using in vivo electrophysiological and behavioral assays in rats. Behavioral experiments showed that Asenapine had no significant effect on immobility time in the forced swim test (FST) in control rats. In the ACTH-treated rats, a model of antidepressant-resistance, Asenapine failed to alter immobility time in the FST. In contrast in the sleep deprivation (SD) model of mania, acute administration of Asenapine significantly decreased the hyperlocomotion of SD rats. In the DRN, acute administration of Asenapine reduced the suppressant effect of the selective 5-HT These results confirm that Asenapine displays robust antimanic property and effective in vivo antagonistic activity at 5-HT