Merck
Chlorogenic Compounds from Coffee Beans Exert Activity against Respiratory Viruses.
Planta medica (2016-11-03)
Valentina Sinisi, Annelies Stevaert, Federico Berti, Cristina Forzato, Fabio Benedetti, Luciano Navarini, Anita Camps, Leentje Persoons, Kurt Vermeire
ZUSAMMENFASSUNG
Chlorogenic acids are secondary metabolites in diverse plants. Some chlorogenic acids extracted from traditional medicinal plants are known for their healing properties, e.g., against viral infections. Also, green coffee beans are a rich source of chlorogenic acids, with 5-