Merck
  • Retinoic Acid Regulates Immune Responses by Promoting IL-22 and Modulating S100 Proteins in Viral Hepatitis.
Retinoic Acid Regulates Immune Responses by Promoting IL-22 and Modulating S100 Proteins in Viral Hepatitis.
Journal of immunology (Baltimore, Md. : 1950) (2017-04-02)
Zuliang Jie, Yuejin Liang, Panpan Yi, Hui Tang, Lynn Soong, Yingzi Cong, Kangling Zhang, Jiaren Sun
ZUSAMMENFASSUNG
Although large amounts of vitamin A and its metabolite all-
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human S100A4