Merck
Rapid Synthesis of Redox-Active Dodecaborane B
Inorganic chemistry frontiers (2016-11-26)
Alex I Wixtrom, Yanwu Shao, Dahee Jung, Charles W Machan, Shaunt N Kevork, Elaine A Qian, Jonathan C Axtell, Saeed I Khan, Clifford P Kubiak, Alexander M Spokoyny
ZUSAMMENFASSUNG
We have developed a fast and efficient route to obtain perfunctionalized ether-linked alkyl and benzyl derivatives of the