Merck
Further evidence of the involvement of the Wnt signaling pathway in Dupuytren's disease.
Journal of cell communication and signaling (2015-12-05)
Evert-Jan P M Ten Dam, Marike M van Beuge, Ruud A Bank, Paul M N Werker
ZUSAMMENFASSUNG
Genetic background plays an important role in the development of Dupuytren's disease. A genome-wide association study (GWAS) showed that nine loci are associated with the disease, six of which contain genes that are involved in Wnt signaling (WNT2, WNT4, WNT7B, RSPO2, SFRP4, SULF1). To obtain insight in the role of these genes, we performed expression studies on affected and unaffected patient's tissues. Surgically obtained nodules and cords from eight Dupuytren's patients were compared to patient-matched control tissue (unaffected transverse palmar fascia). The Wnt-related genes found in the GWAS, the classical Wnt-downstream protein β-catenin, as well as (myo)fibroblast markers were analyzed using real-time qPCR and immunohistochemical stainings for mRNA levels and protein levels, respectively. The collagen-coding genes COL1A1 and COL3A1 were highly upregulated on mRNA level, both in cords and nodules. Three Wnt-related genes were found to be differently regulated compared to control tissue: WNT2 was downregulated in nodules, WNT7B was upregulated in nodules, and SFRP4 was upregulated in nodules and cords. Immunohistochemistry revealed significantly less staining of Wnt2 in cords, but significantly more staining for Wnt7b in nodules. There was significantly more staining of α-SMA in nodules and cord and β-catenin in nodules than in control tissue. We found differences in expression, both at mRNA and protein level, in several Wnt-related genes found earlier to be associated with Dupuytren's disease. Of these, Wnt7b was upregulated and found in close association with both α-SMA and β-catenin expressing cells, making it a candidate pro-fibrotic mediator in Dupuytren's disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-SFRP4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.