- An aberrant phase transition of stress granules triggered by misfolded protein and prevented by chaperone function.
An aberrant phase transition of stress granules triggered by misfolded protein and prevented by chaperone function.
The EMBO journal (2017-04-06)
Daniel Mateju, Titus M Franzmann, Avinash Patel, Andrii Kopach, Edgar E Boczek, Shovamayee Maharana, Hyun O Lee, Serena Carra, Anthony A Hyman, Simon Alberti
PMID28377462
ZUSAMMENFASSUNG
Stress granules (SG) are membrane-less compartments involved in regulating mRNAs during stress. Aberrant forms of SGs have been implicated in age-related diseases, such as amyotrophic lateral sclerosis (ALS), but the molecular events triggering their formation are still unknown. Here, we find that misfolded proteins, such as ALS-linked variants of SOD1, specifically accumulate and aggregate within SGs in human cells. This decreases the dynamics of SGs, changes SG composition, and triggers an aberrant liquid-to-solid transition of
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-FUS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution