Merck
Comparative oncogenomics identifies tyrosine kinase FES as a tumor suppressor in melanoma.
The Journal of clinical investigation (2017-05-04)
Michael Olvedy, Julie C Tisserand, Flavie Luciani, Bram Boeckx, Jasper Wouters, Sophie Lopez, Florian Rambow, Sara Aibar, Bernard Thienpont, Jasmine Barra, Corinna Köhler, Enrico Radaelli, Sophie Tartare-Deckert, Stein Aerts, Patrice Dubreuil, Joost J van den Oord, Diether Lambrechts, Paulo De Sepulveda, Jean-Christophe Marine
ZUSAMMENFASSUNG
Identification and functional validation of oncogenic drivers are essential steps toward advancing cancer precision medicine. Here, we have presented a comprehensive analysis of the somatic genomic landscape of the widely used BRAFV600E- and NRASQ61K-driven mouse models of melanoma. By integrating the data with publically available genomic, epigenomic, and transcriptomic information from human clinical samples, we confirmed the importance of several genes and pathways previously implicated in human melanoma, including the tumor-suppressor genes phosphatase and tensin homolog (PTEN), cyclin dependent kinase inhibitor 2A (CDKN2A), LKB1, and others. Importantly, this approach also identified additional putative melanoma drivers with prognostic and therapeutic relevance. Surprisingly, one of these genes encodes the tyrosine kinase FES. Whereas FES is highly expressed in normal human melanocytes, FES expression is strongly decreased in over 30% of human melanomas. This downregulation correlates with poor overall survival. Correspondingly, engineered deletion of Fes accelerated tumor progression in a BRAFV600E-driven mouse model of melanoma. Together, these data implicate FES as a driver of melanoma progression and demonstrate the potential of cross-species oncogenomic approaches combined with mouse modeling to uncover impactful mutations and oncogenic driver alleles with clinical importance in the treatment of human cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-FES antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution