Merck
  • Molecular and biochemical characterization of recombinant cel12B, cel8C, and peh28 overexpressed in
Molecular and biochemical characterization of recombinant cel12B, cel8C, and peh28 overexpressed in
Biotechnology for biofuels (2017-04-18)
Eman Ibrahim, Kim D Jones, Keith E Taylor, Ebtesam N Hosseney, Patrick L Mills, Jean M Escudero
ZUSAMMENFASSUNG
The high crystallinity of cellulosic biomass myofibrils as well as the complexity of their intermolecular structure is a significant impediment for biofuel production. Cloning of β-Jelly roll topology, (α/α) This study provides some insight into which parameters should be optimized when application of cel8C, cel12B, and peh28 to biomass conversion is the goal.