- Sono-synthesis of solar light responsive S-N-C-tri doped TiO
Sono-synthesis of solar light responsive S-N-C-tri doped TiO
Ultrasonics sonochemistry (2017-06-22)
Sara Ramandi, Mohammad H Entezari, Narjes Ghows
PMID28633823
ZUSAMMENFASSUNG
C-N-S-tri doped TiO