Merck
  • Loss of Calreticulin Uncovers a Critical Role for Calcium in Regulating Cellular Lipid Homeostasis.
Loss of Calreticulin Uncovers a Critical Role for Calcium in Regulating Cellular Lipid Homeostasis.
Scientific reports (2017-07-21)
Wen-An Wang, Wen-Xin Liu, Serpen Durnaoglu, Sun-Kyung Lee, Jihong Lian, Richard Lehner, Joohong Ahnn, Luis B Agellon, Marek Michalak
ZUSAMMENFASSUNG
A direct link between Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin (wasserlöslich), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Lipoprotein-Mangelserum aus fetal calf, frozen liquid
Sigma-Aldrich
MISSION® esiRNA, targeting human MBTPS1