Merck
  • Transcriptome and Metabolite Changes during Hydrogen Cyanamide-Induced Floral Bud Break in Sweet Cherry.
Transcriptome and Metabolite Changes during Hydrogen Cyanamide-Induced Floral Bud Break in Sweet Cherry.
Frontiers in plant science (2017-08-05)
Irina A Ionescu, Gregorio López-Ortega, Meike Burow, Almudena Bayo-Canha, Alexander Junge, Oliver Gericke, Birger L Møller, Raquel Sánchez-Pérez
ZUSAMMENFASSUNG
Release of bud dormancy in perennial woody plants is a temperature-dependent process and thus flowering in these species is heavily affected by climate change. The lack of cold winters in temperate growing regions often results in reduced flowering and low fruit yields. This is likely to decrease the availability of fruits and nuts of the
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Katalase aus Rinderleber, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
α-Hydroxyisobutyronitril-β-D-glucopyranosid, ≥97% (HPLC)