Merck
  • mTOR controls ChREBP transcriptional activity and pancreatic β cell survival under diabetic stress.
mTOR controls ChREBP transcriptional activity and pancreatic β cell survival under diabetic stress.
The Journal of cell biology (2017-06-14)
Gia Cac Chau, Dong Uk Im, Tong Mook Kang, Jeong Mo Bae, Won Kim, Suhkneung Pyo, Eun-Yi Moon, Sung Hee Um
ZUSAMMENFASSUNG
Impaired nutrient sensing and dysregulated glucose homeostasis are common in diabetes. However, how nutrient-sensitive signaling components control glucose homeostasis and β cell survival under diabetic stress is not well understood. Here, we show that mice lacking the core nutrient-sensitive signaling component mammalian target of rapamycin (mTOR) in β cells exhibit reduced β cell mass and smaller islets. mTOR deficiency leads to a severe reduction in β cell survival and increased mitochondrial oxidative stress in chemical-induced diabetes. Mechanistically, we find that mTOR associates with the carbohydrate-response element-binding protein (ChREBP)-Max-like protein complex and inhibits its transcriptional activity, leading to decreased expression of thioredoxin-interacting protein (TXNIP), a potent inducer of β cell death and oxidative stress. Consistent with this, the levels of TXNIP and ChREBP were highly elevated in human diabetic islets and