Merck
  • A Nanoparticle-Based Ophthalmic Formulation of Dexamethasone Enhances Corneal Permeability of the Drug and Prolongs Its Corneal Residence Time.
A Nanoparticle-Based Ophthalmic Formulation of Dexamethasone Enhances Corneal Permeability of the Drug and Prolongs Its Corneal Residence Time.
Biological & pharmaceutical bulletin (2017-07-05)
Noriaki Nagai, Yosuke Nakazawa, Yoshimasa Ito, Kazutaka Kanai, Norio Okamoto, Yoshikazu Shimomura
ZUSAMMENFASSUNG
We designed ophthalmic formulations containing dexamethasone-loaded solid nanoparticles (DEX
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)