Merck
N-type Ca
Neurobiology of aging (2017-04-10)
Flavia R Silva, Artur S Miranda, Rebeca P M Santos, Isabella G Olmo, Gerald W Zamponi, Tomas Dobransky, Jader S Cruz, Luciene B Vieira, Fabiola M Ribeiro
ZUSAMMENFASSUNG
Huntington's disease (HD) is an autosomal dominant neurodegenerative disorder caused by a polyglutamine expansion in the amino-terminal region of the huntingtin (htt) protein. In addition to facilitating neurodegeneration, mutant htt is implicated in HD-related alterations of neurotransmission. Previous data showed that htt can modulate N-type voltage-gated Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade