Merck
Three new drimane sesquiterpenoids from cultures of the fungus Penicillium sp.
Journal of Asian natural products research (2016-11-29)
Jian-Hai Ding, Zhang-Gui Ding, Wei-Xun Chunyu, Jiang-Yuan Zhao, Hai-Bin Wang, Shi-Wei Liu, Fei Wang
ZUSAMMENFASSUNG
Three new drimane sesquiterpenoids, 12-hydroxyalbrassitriol (1), drim-8(12)-en-6β,7α, 9α,11-tetraol (2), and drim-68(12)-dien-9α,11-diol (3), along with one known analog albrassitriol (4), were isolated from cultures of the tin mine tailings-associated fungus Penicillium sp. The new structures were determined on the basis of extensive spectroscopic analyses. All compounds were tested for their cytotoxicities against five human cancer cell lines.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Propylene sulfite, 99%