Merck
  • Small Molecule Mixture Analysis by Heteronuclear NMR under Spin Diffusion Conditions in Viscous DMSO-Water Solvent.
Small Molecule Mixture Analysis by Heteronuclear NMR under Spin Diffusion Conditions in Viscous DMSO-Water Solvent.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-02-15)
Pedro Lameiras, Solène Patis, Jouda Jakhlal, Stéphanie Castex, Pascale Clivio, Jean-Marc Nuzillard
ZUSAMMENFASSUNG
Spin diffusion in NMR occurs for small- and medium-sized molecules when their tumbling rate reduces in solution so that magnetization exchange by longitudinal cross relaxation becomes highly efficient. Composite DMSO-water viscous solvents were used for the first time to access the individual NMR spectra of a mixture components in spin diffusion conditions. The easy handling and high dissolution power of [D
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ala-Tyr