Merck
MicroRNA-138 inhibits cell proliferation in hepatocellular carcinoma by targeting Sirt1.
Oncology reports (2017-07-06)
Jia Luo, Pan Chen, Wei Xie, Feiyue Wu
ZUSAMMENFASSUNG
MicroRNAs (miRNAs) are a family of small, non‑coding RNA molecules that are highly conserved across species and function as regulators of gene expression. In the present study, we revealed that miR-138 expression was at a low level while sirtuin type 1 (Sirt1) mRNA expression was at high level in hepatocellular carcinoma tissues and cell lines by using real-time PCR and western blot assays, and the functions of miR-138 were achieved via targeting of Sirt1 using luciferase reporter gene vector and RNA immunoprecipitation assays. Overexpression of miR-138 attenuated Sirt1 expression and inhibited cell proliferation by using CCK-8 and BrdU assays. The inhibitory effect of miR-138 could be partially restored by forced expression of Sirt1 in cells. Our data revealed a crucial role and mechanism of miR-138 in the regulation of hepatocellular carcinoma cell growth via the miR-138/Sirt1 axis, and miR-138 could be an important potential target for the clinical management of hepatocellular carcinoma in the future.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Imprint® RNA Immunoprecipitation Kit, High-capacity Protein A magnetic beads for successful RNA Immunoprecipitation,suitable for use with mRNA and microRNA
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder