Merck
  • Integrated preservation and sample clean up procedures for studying water ingestion by recreational swimmers via urinary biomarker determination.
Integrated preservation and sample clean up procedures for studying water ingestion by recreational swimmers via urinary biomarker determination.
Analytica chimica acta (2017-07-25)
Ricardo Cantú, Jody A Shoemaker, Catherine A Kelty, Larry J Wymer, Thomas D Behymer, Alfred P Dufour, Matthew L Magnuson
ZUSAMMENFASSUNG
The use of cyanuric acid as a biomarker for ingestion of swimming pool water may lead to quantitative knowledge of the volume of water ingested during swimming, contributing to a better understanding of disease resulting from ingestion of environmental contaminants. When swimming pool water containing chlorinated cyanurates is inadvertently ingested, cyanuric acid is excreted quantitatively within 24 h as a urinary biomarker of ingestion. Because the volume of water ingested can be quantitatively estimated by calculation from the concentration of cyanuric acid in 24 h urine samples, a procedure for preservation, cleanup, and analysis of cyanuric acid was developed to meet the logistical demands of large scale studies. From a practical stand point, urine collected from swimmers cannot be analyzed immediately, given requirements of sample collection, shipping, handling, etc. Thus, to maintain quality control to allow confidence in the results, it is necessary to preserve the samples in a manner that ensures as quantitative analysis as possible. The preservation and clean-up of cyanuric acid in urine is complicated because typical approaches often are incompatible with the keto-enol tautomerization of cyanuric acid, interfering with cyanuric acid sample preparation, chromatography, and detection. Therefore, this paper presents a novel integration of sample preservation, clean-up, chromatography, and detection to determine cyanuric acid in 24 h urine samples. Fortification of urine with cyanuric acid (0.3-3.0 mg/L) demonstrated accuracy (86-93% recovery) and high reproducibility (RSD < 7%). Holding time studies in unpreserved urine suggested sufficient cyanuric acid stability for sample collection procedures, while longer holding times suggested instability of the unpreserved urine. Preserved urine exhibited a loss of around 0.5% after 22 days at refrigerated storage conditions of 4 °C.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Decyltrimethylammoniumbromid, ≥98.0% (NT)