Merck
  • Interactions between macromolecule-bound antioxidants and Trolox during liposome autoxidation: A multivariate approach.
Interactions between macromolecule-bound antioxidants and Trolox during liposome autoxidation: A multivariate approach.
Food chemistry (2017-08-03)
Ecem Evrim Çelik, Jose Manuel Amigo Rubio, Mogens L Andersen, Vural Gökmen
ZUSAMMENFASSUNG
The interactions between free and macromolecule-bound antioxidants were investigated in order to evaluate their combined effects on the antioxidant environment. Dietary fiber (DF), protein and lipid-bound antioxidants, obtained from whole wheat, soybean and olive oil products, respectively and Trolox were used for this purpose. Experimental studies were carried out in autoxidizing liposome medium by monitoring the development of fluorescent products formed by lipid oxidation. Chemometric methods were used both at experimental design and multivariate data analysis stages. Comparison of the simple addition effects of Trolox and bound antioxidants with measured values on lipid oxidation revealed synergetic interactions for DF and refined olive oil-bound antioxidants, and antagonistic interactions for protein and extra virgin olive oil-bound antioxidants with Trolox. A generalized version of logistic function was successfully used for modelling the oxidation curve of liposomes. Principal component analysis revealed two separate phases of liposome autoxidation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carbonsäure, 97%