Merck
  • CD47 overexpression is associated with decreased neutrophil apoptosis/phagocytosis and poor prognosis in non-small-cell lung cancer patients.
CD47 overexpression is associated with decreased neutrophil apoptosis/phagocytosis and poor prognosis in non-small-cell lung cancer patients.
British journal of cancer (2017-06-21)
Lourdes Barrera, Edgar Montes-Servín, Juan-Manuel Hernandez-Martinez, María de Los Ángeles García-Vicente, Elizabeth Montes-Servín, Marytere Herrera-Martínez, José C Crispín, José Rafael Borbolla-Escoboza, Oscar Arrieta
ZUSAMMENFASSUNG
Non-small-cell lung cancer (NSCLC) patients often exhibit neutrophilia, which has been associated with poor clinical outcomes. However, the mechanisms that lead to neutrophilia have not been fully established. CD47 is an antiphagocytic molecule that promotes neutrophil recruitment. Blood was collected from 50 treatment-naive patients with advanced NSCLC and from 25 healthy subjects. The frequency of CD66b The percentage of circulating neutrophils was significantly higher in patients than in controls (P<0.001). Patient-derived neutrophils had a higher oxidative potential than those of controls (P=0.0286). The number of neutrophils in late apoptosis/necrosis was lower in patients than in controls (P=0.0317). Caspase 3/7 activation was also lower in patients than in controls (P=0.0079). CD47 expression in whole-blood samples and in the neutrophil fraction was higher in NSCLC patients than in controls (P=0.0408 and P<0.001). Patient-derived neutrophils were phagocytosed at a lower rate than those of controls (P=0.0445). CD47 expression in neutrophils negatively correlated with their ingestion by macrophages (P=0.0039). High CD47 expression was associated with a lower overall survival. Increased CD47 expression on the surface of neutrophils was associated with a delay in neutrophil apoptosis and with an impairment in their phagocytic clearance by macrophages, suggesting that CD47 overexpression may be one of the underlying mechanisms leading to neutrophilia in NSCLC patients.