Merck
  • Potential enterotoxicity and antimicrobial resistance pattern of Aeromonas species isolated from pet turtles and their environment.
Potential enterotoxicity and antimicrobial resistance pattern of Aeromonas species isolated from pet turtles and their environment.
The Journal of veterinary medical science (2017-04-11)
S H M P Wimalasena, Gee-Wook Shin, Sabrina Hossain, Gang-Joon Heo
ZUSAMMENFASSUNG
To investigate the potential enterotoxicity and antimicrobial resistance of aeromonads from pet turtles as a risk for human infection, one hundred and two Aeromonas spp. were isolated from the feces, skin and rearing environments of pet turtles and identified by biochemical and gyrB sequence analyses. Aeromonas enteropelogenes was the predominant species among the isolates (52.9%) followed by A. hydrophila (32.4%), A. dharkensis (5.9%), A. veronii (4.9%) and A. caviae (3.9%). Their potential enterotoxicities were evaluated by PCR assays for detecting genes encoding cytotoxic enterotoxin (act) and two cytotonic enterotoxins (alt and ast). 75.8% of A. hydrophila isolates exhibited the act
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Imipenem, Pharmaceutical Secondary Standard; Certified Reference Material