Merck
Micropellets coated with Kollicoat
Drug development and industrial pharmacy (2017-05-10)
Andriy Dashevskiy, Valentyn Mohylyuk, Abid Riaz Ahmed, Karl Kolter, Felicitas Guth, Roland Bodmeier
ZUSAMMENFASSUNG
The objective of this study was to develop delivery systems for taste masking based on multiparticulates coated with Kollicoat
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-(Diethylamino)ethylmethacrylat, contains 1500 ppm MEHQ as inhibitor, 99%