Merck
  • Effect of MPTP on Serotonergic Neuronal Systems and Mitochondrial Complex I Activity in the Living Brain: A PET Study on Conscious Rhesus Monkeys.
Effect of MPTP on Serotonergic Neuronal Systems and Mitochondrial Complex I Activity in the Living Brain: A PET Study on Conscious Rhesus Monkeys.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2017-03-11)
Masakatsu Kanazawa, Hiroyuki Ohba, Shingo Nishiyama, Takeharu Kakiuchi, Hideo Tsukada
ZUSAMMENFASSUNG
The objective of the present PET study was to compare the effect of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) on serotonergic neuronal systems and mitochondrial complex I (MC-I) activity with that of dopamine in conscious rhesus monkeys (
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tri(o-tolyl)phosphin, 97%