Merck
  • A defined metabolic state in pre B cells governs B-cell development and is counterbalanced by Swiprosin-2/EFhd1.
A defined metabolic state in pre B cells governs B-cell development and is counterbalanced by Swiprosin-2/EFhd1.
Cell death and differentiation (2017-05-20)
Merle Stein, Sebastian Dütting, Dimitrios Mougiakakos, Michael Bösl, Kristin Fritsch, Dorothea Reimer, Sophia Urbanczyk, Tobit Steinmetz, Wolfgang Schuh, Aline Bozec, Thomas H Winkler, Hans-Martin Jäck, Dirk Mielenz
ZUSAMMENFASSUNG
B-cell development in the bone marrow comprises proliferative and resting phases in different niches. We asked whether B-cell metabolism relates to these changes. Compared to pro B and small pre B cells, large pre B cells revealed the highest glucose uptake and ROS but not mitochondrial mass, whereas small pre B cells exhibited the lowest mitochondrial membrane potential. Small pre B cells from Rag1
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carbonylcyanid-3-chlorphenylhydrazon, ≥97% (TLC), powder
Sigma-Aldrich
Tetramethylrhodamin-methylester-perchlorat, ≥95%