Merck
Nano-Calorimetry based point of care biosensor for metabolic disease management.
Biomedical microdevices (2017-06-01)
Evan Kazura, Brad R Lubbers, Elliott Dawson, John A Phillips, Franz Baudenbacher
ZUSAMMENFASSUNG
Point of care (POC) diagnostics represents one of the fastest growing health care technology segments. Developments in microfabrication have led to the development of highly-sensitive nanocalorimeters ideal for directly measuring heat generated in POC biosensors. Here we present a novel nano-calorimeter-based biosensor design with differential sensing to eliminate common mode noise and capillary microfluidic channels for sample delivery to the thermoelectric sensor. The calorimeter has a resolution of 1.4 ± 0.2 nJ/(Hz)
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Katalase aus Rinderleber, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)