- Sorption mechanism of enrofloxacin on humic acids extracted from Brazilian soils.
Sorption mechanism of enrofloxacin on humic acids extracted from Brazilian soils.
Environmental science and pollution research international (2017-05-26)
Mónica J Martínez-Mejía, Isabela Sato, Susanne Rath
PMID28537016
ZUSAMMENFASSUNG
Veterinary antimicrobials are emerging environmental contaminants of concern. In this study, the sorption of enrofloxacin (ENR) onto humic acids (HAs) extracted from three Brazilian soils was evaluated. HAs were characterized by elemental analysis and solid
MATERIALIEN