Merck
  • The Bile Acid Receptor GPBAR1 Regulates the M1/M2 Phenotype of Intestinal Macrophages and Activation of GPBAR1 Rescues Mice from Murine Colitis.
The Bile Acid Receptor GPBAR1 Regulates the M1/M2 Phenotype of Intestinal Macrophages and Activation of GPBAR1 Rescues Mice from Murine Colitis.
Journal of immunology (Baltimore, Md. : 1950) (2017-06-14)
Michele Biagioli, Adriana Carino, Sabrina Cipriani, Daniela Francisci, Silvia Marchianò, Paolo Scarpelli, Daniele Sorcini, Angela Zampella, Stefano Fiorucci
ZUSAMMENFASSUNG
GPBAR1 (TGR5 or M-BAR) is a G protein-coupled receptor for secondary bile acids that is highly expressed in monocytes/macrophages. In this study, we aimed to determine the role of GPBAR1 in mediating leukocyte trafficking in chemically induced models of colitis and investigate the therapeutic potential of BAR501, a small molecule agonist for GPBAR1. These studies demonstrated that GPBAR1 gene ablation enhanced the recruitment of classically activated macrophages in the colonic lamina propria and worsened the severity of inflammation. In contrast, GPBAR1 activation by BAR501 reversed intestinal inflammation in the trinitrobenzenesulfonic acid and oxazolone models by reducing the trafficking of Ly6C
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, purified by phenol extraction