Merck
Saudi pharmaceutical journal : SPJ : the official publication of the Saudi Pharmaceutical Society (2017-07-21)
Betul Demirci, Hasan Soliman Yusufoglu, Nurhayat Tabanca, Halide Edip Temel, Ulrich R Bernier, Natasha M Agramonte, Saleh Ibrahim Alqasoumi, Adnan Jathlan Al-Rehaily, Kemal Husnu Can Başer, Fatih Demirci
ZUSAMMENFASSUNG
The essential oil (EO) of the aerial parts of
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Diethyl-3-Methylbenzamid, 97%