Merck
  • Synthesis, characterization of amide substituted dexibuprofen derivatives and their spectral, voltammetric and docking investigations for DNA binding interactions.
Synthesis, characterization of amide substituted dexibuprofen derivatives and their spectral, voltammetric and docking investigations for DNA binding interactions.
Journal of photochemistry and photobiology. B, Biology (2017-03-21)
Nasima Arshad, Muhammad Zafran, Zaman Ashraf, Fouzia Perveen
ZUSAMMENFASSUNG
Three amide derivatives - methyl-2-[2-(4-isobutylphenyl)propanamido]propanoate (Dex-2), methyl 2-[2-(4-isobutylphenyl) propanamido]-3-phenylpropanoate (Dex-3) and methyl 2-[2-(4-isobutylphenyl)-propanamido]-4-methylpentanoate (Dex-4) of dexibuprofen (Dex-1) 2-(4-isobutylphenyl)propanoic acid were synthesized and conformed for structures by physical data and spectral analysis. Further, all the compounds were studied for their binding with ds.DNA through experimental (UV-visible/and fluorescence spectroscopy, cyclic voltammetry) and theoretical (molecular docking) techniques. Spectral and voltammetric responses as well as kinetic and thermodynamic data interpretations at stomach (4.7) and blood (7.4) pH and at human body temperature (37°C) indicated spontaneous interaction of all the compounds with DNA via intercalation and external bindings. The binding constants (K
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%