Merck
Silencing of NRAGE induces autophagy via AMPK/Ulk1/Atg13 signaling pathway in NSCLC cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2017-06-24)
Yiyang Zhou, Nan Huang, Jianchun Wu, Ni Zhen, Ning Li, Yan Li, Yong-Xin Li
ZUSAMMENFASSUNG
NRAGE has been reported to be overexpressed in cancer cells, especially in lung cancer cells. To determine the role of NRAGE in non-small-cell lung cancer cells, we investigated the effects of NRAGE on autophagy in non-small-cell lung cancer cells. Human A549 and H1299 cells were transfected with NRAGE-specific small-interfering RNA. The Cell Counting Kit-8 and plate clone assay showed that downregulation of NRAGE could induce the proliferation in A549 and H1299 cells. In addition, our data suggested that downregulation of NRAGE enhances autophagosome formation by immunofluorescence. We found that knockdown of NRAGE induced autophagy, together with downregulation of P62 and upregulation of LC3-II protein. Furthermore, to elucidate the mechanism of NRAGE in suppressing autophagy, the protein expressions of AMPK, Ulk1, and Atg13 were assessed. Collectively, these results demonstrate the effective anti-autophagic of NRAGE in non-small-cell lung cancer cells through AMPK/Ulk1/Atg13 autophagy signaling pathways. Therefore, NRAGE could be used as a potential therapeutic target for lung cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder