Merck
  • Chronic treatment with a smart antioxidative nanoparticle for inhibition of amyloid plaque propagation in Tg2576 mouse model of Alzheimer's disease.
Chronic treatment with a smart antioxidative nanoparticle for inhibition of amyloid plaque propagation in Tg2576 mouse model of Alzheimer's disease.
Scientific reports (2017-06-21)
Phetcharat Boonruamkaew, Pennapa Chonpathompikunlert, Long Binh Vong, Sho Sakaue, Yasushi Tomidokoro, Kazuhiro Ishii, Akira Tamaoka, Yukio Nagasaki
ZUSAMMENFASSUNG
The present study aimed to assess whether our newly developed redox nanoparticle (RNP