Merck
Primary cilia regulate the osmotic stress response of renal epithelial cells through TRPM3.
American journal of physiology. Renal physiology (2017-01-27)
Brian J Siroky, Nancy K Kleene, Steven J Kleene, Charles D Varnell, Raven G Comer, Jialiu Liu, Lu Lu, Nolan W Pachciarz, John J Bissler, Bradley P Dixon
ZUSAMMENFASSUNG
Primary cilia sense environmental conditions, including osmolality, but whether cilia participate in the osmotic response in renal epithelial cells is not known. The transient receptor potential (TRP) channels TRPV4 and TRPM3 are osmoresponsive. TRPV4 localizes to cilia in certain cell types, while renal subcellular localization of TRPM3 is not known. We hypothesized that primary cilia are required for maximal activation of the osmotic response of renal epithelial cells and that ciliary TRPM3 and TRPV4 mediate that response. Ciliated [murine epithelial cells from the renal inner medullary collecting duct (mIMCD-3) and 176-5] and nonciliated (176-5Δ) renal cells expressed
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-Tubulin, acetyliert in Maus hergestellte Antikörper, clone 6-11B-1, ascites fluid
Sigma-Aldrich
2-Phenylindol, technical grade, 95%