Merck
  • Ginkgolide K attenuates neuronal injury after ischemic stroke by inhibiting mitochondrial fission and GSK-3β-dependent increases in mitochondrial membrane permeability.
Ginkgolide K attenuates neuronal injury after ischemic stroke by inhibiting mitochondrial fission and GSK-3β-dependent increases in mitochondrial membrane permeability.
Oncotarget (2017-06-08)
Xu Zhou, Hui-Ying Wang, Bin Wu, Cai-Yi Cheng, Wei Xiao, Zhen-Zhong Wang, Yu-Yu Yang, Ping Li, Hua Yang
ZUSAMMENFASSUNG
Ginkgolide K (GK) belongs to the ginkgolide family of natural compounds found in Ginkgo biloba leaves, which have been used for centuries to treat cerebrovascular and cardiovascular diseases. We evaluated the protective effects of GK against neuronal apoptosis by assessing its ability to sustain mitochondrial integrity and function. Co-immunoprecipitation showed that Drp1 binding to GSK-3β was increased after an oxygen-glucose deprivation/reperfusion (OGD/R) insult in cultured neuroblastoma cells. This induced Drp1 and GSK-3β translocation to mitochondria and mitochondrial dysfunction, which was attenuated by GK. GK also reduced mitochondrial fission by increasing Drp1 phosphorylation at Ser637 and inhibiting mitochondrial Drp1 recruitment. In addition, GK exposure induced GSK-3β phosphorylation at Ser9 and enhanced the interaction between adenine nucleotide translocator (ANT) and p-GSK-3β. This interaction suppressed the interaction between ANT and cyclophilin D (CypD), which inhibited mitochondrial permeability transition pore (mPTP) opening. Similarly, suppression of mitochondrial fission by Mdivi-1 also inhibited GSK-3β-induced mPTP opening. Treating mice with GK prevented GSK-3β and Drp1 translocation to mitochondria and attenuated mitochondrial dysfunction after middle cerebral artery occlusion. We therefore propose that by inhibiting mitochondrial fission and attenuating mPTP opening, GK exerts neuroprotective effects that mitigate or prevent neuronal damage secondary to ischemic stroke.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetramethylrhodamin-Ethylester-Perchlorat, suitable for fluorescence, ≥90% (HPCE)