Merck
Two Glycerol-3-Phosphate Dehydrogenases from
Plant physiology (2017-06-08)
Thomas Driver, Drupad K Trivedi, Owen A McIntosh, Andrew P Dean, Royston Goodacre, Jon K Pittman
ZUSAMMENFASSUNG
The metabolism of glycerol-3-phosphate (G3P) is important for environmental stress responses by eukaryotic microalgae. G3P is an essential precursor for glycerolipid synthesis and the accumulation of triacylglycerol (TAG) in response to nutrient starvation. G3P dehydrogenase (GPDH) mediates G3P synthesis, but the roles of specific GPDH isoforms are currently poorly understood. Of the five GPDH enzymes in the model alga
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-D-lactosamin, ≥98% (TLC)