Merck
  • Expression of TAU in insulin-secreting cells and its interaction with the calcium-binding protein secretagogin.
Expression of TAU in insulin-secreting cells and its interaction with the calcium-binding protein secretagogin.
The Journal of endocrinology (2010-01-12)
Magdalena Maj, Wolfgang Gartner, Aysegul Ilhan, Dashurie Neziri, Johannes Attems, Ludwig Wagner
ZUSAMMENFASSUNG
Tauopathies have been associated with Alzheimer's disease (AD), which frequently manifests together with diabetes mellitus type 2. Calcium-binding proteins such as the recently identified secretagogin (SCGN) might exert protective effects. As pancreatic beta-cells and neurons share common electrophysiological properties, we investigated the appearance of TAU (listed as MAPT in the HUGO and MGI Databases) protein at the islets of Langerhans and beta-cell-derived cell lines which highly express the neuroendocrine-specific protein SCGN. Six predominant TAU isoforms could be identified by immunoblotting, which formed TAU deposits detectable by immunofluorescence and sarkosyl-insoluble pellets. Using GST-SCGN pull-down assays, a calcium-dependent SCGN-TAU interaction was found. In this line, sucrose density gradient fractionation and differential ultracentrifugation studies of TAU and SCGN revealed co-appearance of both proteins. Co-localization of TAU and SCGN within insulinoma cells and islets of Langerhans mainly restricted to insulin-positive beta-cells was demonstrated by confocal microscopy. Motivated by these findings, we looked if SCGN overexpression could exert protective function on Rin-5F cells, which showed differences in TAU levels. Testing the vulnerability of Rin-5F clones by MTT assay, we revealed that high TAU levels going along with highest TAU aggregates could not be antagonized by high levels of SCGN protein. Our findings demonstrated for the first time the association of TAU and the calcium-binding protein SCGN and support earlier results implicating that beta-cells might represent an extra cerebral site of tauopathy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Tau (3-Repeat Isoform RD3)-Antikörper, Klon 8E6/C11, culture supernatant, clone 8E6/C11, Upstate®