Merck
Nano-sized cytochrome P450 3A4 inhibitors to block hepatic metabolism of docetaxel.
International journal of nanomedicine (2017-08-18)
Marion Paolini, Laurence Poul, Céline Berjaud, Matthieu Germain, Audrey Darmon, Maxime Bergère, Agnès Pottier, Laurent Levy, Eric Vibert
ZUSAMMENFASSUNG
Most drugs are metabolized by hepatic cytochrome P450 3A4 (CYP3A4), resulting in their reduced bioavailability. In this study, we present the design and evaluation of bio-compatible nanocarriers trapping a natural CYP3A4-inhibiting compound. Our aim in using nanocarriers was to target the natural CYP3A4-inhibiting agent to hepatic CYP3A4 and leave drug-metabolizing enzymes in other organs undisturbed. In the design of such nanocarriers, we took advantage of the nonspecific accumulation of small nanoparticles in the liver. Specific targeting functionalization was added to direct nanocarriers toward hepatocytes. Nanocarriers were evaluated in vitro for their CYP3A4 inhibition capacity and in vivo for their biodistribution, and finally injected 24 hours prior to the drug docetaxel, for their ability to improve the efficiency of the drug docetaxel. Nanoparticles of poly(lactic-
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-D-galactosamin, ≥98% (HPLC)