Merck
  • bioA mutant of Mycobacterium tuberculosis shows severe growth defect and imparts protection against tuberculosis in guinea pigs.
bioA mutant of Mycobacterium tuberculosis shows severe growth defect and imparts protection against tuberculosis in guinea pigs.
PloS one (2017-06-29)
Ritika Kar, Prachi Nangpal, Shubhita Mathur, Swati Singh, Anil K Tyagi
ZUSAMMENFASSUNG
Owing to the devastation caused by tuberculosis along with the unsatisfactory performance of the Bacillus Calmette-Guérin (BCG) vaccine, a more efficient vaccine than BCG is required for the global control of tuberculosis. A number of studies have demonstrated an essential role of biotin biosynthesis in the growth and survival of several microorganisms, including mycobacteria, through deletion of the genes involved in de novo biotin biosynthesis. In this study, we demonstrate that a bioA mutant of Mycobacterium tuberculosis (MtbΔbioA) is highly attenuated in the guinea pig model of tuberculosis when administered aerogenically as well as intradermally. Immunization with MtbΔbioA conferred significant protection in guinea pigs against an aerosol challenge with virulent M. tuberculosis, when compared with the unvaccinated animals. Booster immunization with MtbΔbioA offered no advantage over a single immunization. These experiments demonstrate the vaccinogenic potential of the attenuated M. tuberculosis bioA mutant against tuberculosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridoxin, ≥98%
Sigma-Aldrich
Ammoniumcitrat tribasisch, ≥97% (titration)
Sigma-Aldrich
Thiophen-2-carbonsäurehydrazid, ≥98%