- Highly efficient hydrogen evolution reaction by strain and phase engineering in composites of Pt and MoS
Highly efficient hydrogen evolution reaction by strain and phase engineering in composites of Pt and MoS
Physical chemistry chemical physics : PCCP (2017-07-06)
Da Young Hwang, Kyoung Hwan Choi, Jeong Eon Park, Dong Hack Suh
PMID28678277
ZUSAMMENFASSUNG
The phase transition through local strain engineering is an exciting avenue for controlling electronic, magnetic properties and catalyst activity of materials but complex phenomenon in nanoscience. Herein, we demonstrate the first combinations of bending strain and 2H/1T phase transition by rolling up MoS